Skip to Content
Merck
All Photos(1)

Key Documents

8.00510

Sigma-Aldrich

Palmitoyl chloride

for synthesis

Synonym(s):

Palmitoyl chloride, Hexadecanoyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)14COCl
CAS Number:
Molecular Weight:
274.87
MDL number:
UNSPSC Code:
12352106
EC Index Number:
203-996-7
NACRES:
NA.22

Quality Level

Assay

≥98.0% (GC)

form

liquid

potency

>2000 mg/kg LD50, oral (Rat)

bp

199 °C/27 hPa

mp

12 °C

transition temp

flash point 182 °C

density

0.91 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

ClC(=O)CCCCCCCCCCCCCCC

InChI

1S/C16H31ClO/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3

InChI key

ARBOVOVUTSQWSS-UHFFFAOYSA-N

Related Categories

Application



Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.906 - 0.912
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

320.0 °F - closed cup

Flash Point(C)

160 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service