Skip to Content
Merck
All Photos(1)

Key Documents

574775-M

Sigma-Aldrich

Swainsonine, Swainsona canescens

Reversible active-site inhibitor of lysosomal α-mannosidase.

Synonym(s):

Swainsonine, Swainsona canescens, (1S,2S,8R,8aR)-Trihydroxyindolizidine, 8aβ-Indolizidine-1α,2α,8β-triol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15NO3
CAS Number:
Molecular Weight:
173.21
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 1 mg/mL
DMSO: 5 mg/mL
chloroform: soluble

storage temp.

2-8°C

SMILES string

N21[C@@H]([C@@H]([C@@H](C2)O)O)[C@@H](CCC1)O

InChI

1S/C8H15NO3/c10-5-2-1-3-9-4-6(11)8(12)7(5)9/h5-8,10-12H,1-4H2/t5-,6-,7-,8-/m1/s1

InChI key

FXUAIOOAOAVCGD-WCTZXXKLSA-N

General description

Reversible active-site inhibitor of lysosomal α-mannosidase. Inhibits the growth of tumors and prevents metastasis in murine models. Blocks the processing of high mannose to complex type oligosaccharides.

Biochem/physiol Actions

Primary Target
Lysosomal α-mannosidase

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Mochizuki, H., et al. 1993. J. Biol. Chem. 268, 9143.
Elbein, A.D. 1983. Methods Enzymol.98, 135.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service