Skip to Content
Merck
All Photos(1)

Key Documents

565767

Sigma-Aldrich

α-Secretase Substrate II, Fluorogenic

An internally quenched fluorogenic peptide substrate containing the α-secretase cleavage site of β-Amyloid Precursor Protein (APP).

Synonym(s):

α-Secretase Substrate II, Fluorogenic, Ac-RE(EDANS)-VHHQKLVF-K(DABCYL)-R-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C100H144N30O20S
Molecular Weight:
2118.47
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

red

solubility

DMSO: 5 mg/mL

fluorescence

λex ~340 nm
λem ~490 nm

shipped in

ambient

storage temp.

−20°C

General description

An internally quenched fluorogenic peptide substrate that contains the α-secretase cleavage site of β-amyloid precursor protein (APP). Shown to be an effective substrate for yapsin-2, a glycosyl-phosphatidylinositol-linked aspartyl protease found in yeast (Vmax/Km = 0.031 min-1). The cleavage has been shown to occur at the Lys-Leu bond. Excitation max: ~340 nm; emission max: ~490 nm.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Substrate containing the α-secretase cleavage site of β-Amyloid Precursor Protein (APP)
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

Ac-Arg-Glu(EDANS)-Val-His-His-Gln-Lys-Leu-Val-Phe-Lys(DABCYL)-Arg-OH

Other Notes

Komano, H., et al. 1999. J. Biol. Chem. 274, 24431.
Komano, H., et al. 1998. J. Biol. Chem. 273, 31648.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service