Skip to Content
Merck
All Photos(1)

Key Documents

539644

Sigma-Aldrich

UCN-01

A cell-permeable Staurosporine derived anticancer agent that reversibly and ATP-competitively inhibits several protein kinases.

Synonym(s):

UCN-01, 7-Hydroxystaurosporine, Staurosporine, 7β-Hydroxy, Streptomyces sp., Chk1 Inhibitor VI, Chk2 Inhibitor V

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H26N4O4
CAS Number:
Molecular Weight:
482.53
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

tan
white to beige

solubility

ethanol: 1 mg/mL
DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[n]21c3c4[n](c6c(c4c7c(c3c8c2cccc8)[C@H](NC7=O)O)cccc6)[C@@H]5O[C@]1([C@@H]([C@@H](C5)NC)OC)C

InChI

1S/C28H26N4O4/c1-28-25(35-3)15(29-2)12-18(36-28)31-16-10-6-4-8-13(16)19-21-22(27(34)30-26(21)33)20-14-9-5-7-11-17(14)32(28)24(20)23(19)31/h4-11,15,18,25,27,29,34H,12H2,1-3H3,(H,30,33)/t15-,18-,25-,27-,28+/m1/s1

InChI key

PBCZSGKMGDDXIJ-HQCWYSJUSA-N

General description

A cell-permeable Staurosporine (Cat. No. 569397) derived anticancer agent that reversibly and ATP-competitively inhibits several protein kinases (IC50 = 29 nM, 34 nM, 30 nM, 590 nM and 530 nM for PKCα, PKCβ, PKCγ, PKCδ and PKCε; IC50 = 7 nM, 27 nM, 50 nM, 50 nM, 150 nM and 1.04 M for Chk1, Cdc25C-associated protein kinase 1, Cdk1, PAK4, Cdk5/p25 and Chk2; IC50 = 33 nM, 50 nM, 95 nM, 500 nM, 500 nM and 1.0 M for PDK1, lck, MAPKAP kinase-2, Akt, GSK-3β and PKA, respectively). At higher concentrations (>15 M), affects the activities of Src, PIM-1, CKII, DNA-PK, ErK1, ILK-1 and MAPKK1). Reported to suppress thymidylate synthase expression, induce apoptosis with caspase activation, and sensitize tumor cells to a range of DNA-damaging agents.

Biochem/physiol Actions

Cell permeable: yes
IC50 = 29 nM, 34 nM, 30 nM, 590 nM, 530 nM, 7 nM, 27 nM, 50 nM, 50 nM, 150 nM, 1.04 µM, 33 nM, 50 nM, 95 nM, 500 nM, 500 nM and 1.0 µM, respectively for primary targets in the order listed
Primary Target
PKCα, PKCβ, PKCγ, PKCδ PKCε, Chk1, Cdc25C-associated protein kinase 1, Cdk1, PAK4, Cdk5/p25 and Chk2, PDK1, lck, MAPKAP kinase-2, Akt, GSK-3β and PKA
Product competes with ATP.
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Reinhardt, H.C., et al. 2007. Cancer Cell11, 175.
Jiang, X., et al. 2004. Mol. Cancer Ther.3, 1221.
Sato, S., et al. 2002. Oncogene21, 1727.
Patel, V., et al. 2002. Clin. Cancer Res.8, 3549.
Leclerc, S., et al. 2001. J. Biol. Chem.276, 251.
Busby, E.C., et al. 2000. Cancer Res.60, 2108.
Hsueh, C.T., et al. 1998. Clin. Cancer Res.4, 2201.
Akiyama, T., et al. 1997. Cancer Res.57, 1495.
Seynaeve, C.M., et al. 1994. Mol. Pharmacol.45, 1207.
Takahashi, I., et al. 1987, J. Antibiot.40, 1782.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bin Peng et al.
Nucleic acids research, 49(13), 7554-7570 (2021-07-02)
Polo-like kinase 1 (PLK1) is a master kinase that regulates cell cycle progression. How its enzymatic activity is regulated in response to DNA damage is not fully understood. We show that PLK1 is enriched at double strand breaks (DSBs) within

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service