Skip to Content
Merck
All Photos(1)

Key Documents

506345

Sigma-Aldrich

Palmitic Acid

Synonym(s):

Palmitic Acid, Hexadecanoic Acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H32O2
CAS Number:
Molecular Weight:
256.42
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥99% (GC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤0.05% sulfated ash

color

white

solubility

hot ethanol: soluble

cation traces

Fe: ≤0.0005%
heavy metals (as Pb): ≤0.001%

shipped in

ambient

storage temp.

10-30°C

SMILES string

[O-]C(=O)CCCCCCCCCCCCCCC.[H+]

InChI

1S/C16H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3,(H,17,18)

InChI key

IPCSVZSSVZVIGE-UHFFFAOYSA-N

Biochem/physiol Actions

Cell permeable: no
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

235.4 °F

Flash Point(C)

113 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service