Skip to Content
Merck
All Photos(1)

Key Documents

437692

Sigma-Aldrich

α-Lipoic Acid

α-Lipoic Acid, CAS 1077-28-7 is an oxidized form with antioxidant properties. Reduces renal lipid peroxidation in rats with cisplatin n-induced nephrotoxicity.

Synonym(s):

α-Lipoic Acid, DL-Thioctic Acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14O2S2
CAS Number:
Molecular Weight:
206.33
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.77

Quality Level

description

Merck USA index - 14, 9326

Assay

≥99% (by assay)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

pale yellow

solubility

ethanol: soluble
methanol: soluble

shipped in

ambient

storage temp.

10-30°C

SMILES string

S1SCCC1CCCCC(=O)O

InChI

1S/C8H14O2S2/c9-8(10)4-2-1-3-7-5-6-11-12-7/h7H,1-6H2,(H,9,10)

InChI key

AGBQKNBQESQNJD-UHFFFAOYSA-N

General description

Oxidized form. Has antioxidant properties. Reported to effectively reduce renal lipid peroxidation in rats with cisplatin-induced nephrotoxicity. Acts as a growth factor for many bacteria and protozoa.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Reduce renal lipid peroxidation in rats with cisplatin-induced nephrotoxicity
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Harmful (C)

Other Notes

Somami, S. M., et al. 2000. Pharmacol. Toxicol. 86, 234.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service