Skip to Content
Merck
All Photos(1)

Documents

407254

Sigma-Aldrich

IGFBP-3, CF, Mouse, Recombinant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

Assay

≥95% (SDS-PAGE)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

<1.0 EU/μg protein Endotoxin

shipped in

ambient

storage temp.

−20°C

General description

Recombinant, mouse insulin-like growth factor binding protein 3 (IGFBP-3) consisting of amino acids 22-291 and expressed in NS0-derived murine myeloma cells with four substitutions: Arg250Gln, Gln259Arg, Ser260Gly, Arg271Pro. Mouse IGFBP-3 cDNA encodes a 291 amino acid residue precursor protein with a putative 27 aa residue signal peptide.

Biochem/physiol Actions

ED₅₀ = 0.125-0.5 µg/ml as measured by inhibition of the biological activity of IGF-I or IGF-II (30 ng/ml) on MCF-7 human breast adenocarcinoma cells.

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 0.2 µm filtered Acetonitrile, TFA.

Reconstitution

Following reconstitution refrigerate (4°C) for short-term storage or aliquot and freeze (-20°C) for long-term storage. Stock solutions are stable for up to 1 month at 4°C or for up to 3 months at -20°C.
Reconstitute to 100 µg/ml in sterile PBS.

Other Notes

Schuller, A.G.P., et al. 1994. Mol. Cell. Endoc.104, 57.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service