Skip to Content
Merck
All Photos(1)

Key Documents

236015

Sigma-Aldrich

COX-2 Inhibitor V, FK3311

The COX-2 Inhibitor V, FK3311, also referenced under CAS 116686-15-8, controls the biological activity of COX-2. This small molecule/inhibitor is primarily used for Cell Signaling applications.

Synonym(s):

COX-2 Inhibitor V, FK3311, 4ʹ-Acetyl-2ʹ-(2,4-Difluorophenoxy)methanesulfonanilide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13F2NO4S
CAS Number:
Molecular Weight:
341.33
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

ethanol: 15 mg/mL
DMSO: 30 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Fc1c(ccc(c1)F)Oc2c(ccc(c2)C(=O)C)N[S](=O)(=O)C

InChI

1S/C15H13F2NO4S/c1-9(19)10-3-5-13(18-23(2,20)21)15(7-10)22-14-6-4-11(16)8-12(14)17/h3-8,18H,1-2H3

InChI key

DIIYLGZNZGPXRR-UHFFFAOYSA-N

General description

A cell-permeable and orally available sulfonanilide that acts as a COX-2-selective non-steroidal anti-inflammatory drug (NSAID). Shown to exhibit 500-fold greater potency in inhibiting the LPS-induced (IC50 = 316 nM in human mononuclear cells) than the non-induced/constitutive (IC50 = 160 µM in human platelets) TxB2 production in vitro and effectively alleviate warm ischemia-reperfusion injury in the canine liver and rat lung in vivo.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Otani, Y., et al. 2007. J. Invest. Surg.20, 175.
Sunose, Y., et al. 2001. Transplant Proc.33, 862.
Grossman, C.J., et al. 1995. Inflamm. Res.44, 253.
Nakamura, K., et al. 1993. Chem. Pharm. Bull.41, 2050.
Tsuji, K., et al. 1992. Chem. Pharm. Bull.40, 2399.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service