Skip to Content
Merck
All Photos(5)

Key Documents

1.40141

Millipore

Buffered Peptone Water

EcoCult®, ISO 6579, ISO 6887, ISO 21528, ISO 22964, FDA-BAM, EP 2.6.31, powder, pack of 500 g or 5 kg or 10 kg

Synonym(s):

EcoCult® Buffered Peptone Water, Tryptone Phosphate Water, Peptone Water (buffered), BPW, Buffered Peptone medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

Agency

APHA
EP 2.6.31
FDA-BAM
ISO 21528
ISO 22964
ISO 6579
ISO 6887

Quality Level

sterility

non-sterile

form

powder

manufacturer/tradename

EcoCult®

packaging

pack of 500 g or 5 kg or 10 kg

technique(s)

microbiological culture: suitable

color

light yellow Powder
clear yellowish Prepared medium

application(s)

food and beverages
microbiology
pathogen testing
pharmaceutical

storage temp.

10-30°C

suitability

Cronobacter spp. (spp.)
Salmonella spp. (spp.)
enterobacteriaceae
nonselective for bacteria (preliminary enrichment)

General description

EcoCult® Buffered Peptone Water is rich in nutrients and produces high resuscitation rates for sub-lethally injured bacteria and intense growth. The phosphate buffer system prevents bacterial damage caused by changes in the pH of the medium. Peptone, including an enzymatic digest of casein, acts as a source of carbon, nitrogen, vitamins, and minerals. Sodium chloride helps to maintain the osmotic balance.

Application

EcoCult® Buffered Peptone Water is used for the preliminary non-selective enrichment of bacteria, particularly pathogenic Enterobacteriaceae such as Salmonella and Cronobacter spp, from food and animal feed, water, and other materials. It is also suitable for the dissolving, suspending, and diluting of test samples from food, animal feed, and other materials.

Features and Benefits

  • Compliance to major regulatory standards ISO, FDA-BAM, EPA or USP
  • Clear labeling and Certificate of Analysis to check compliance at a glance
  • Reliable and accurate quality in powder format
  • Cost-effective pricing

Storage and Stability

  • Store at +10 °C to +30 °C, dry and tightly closed.
  • Do not use clumped or discolored medium.
  • Protect from UV light (including sunlight).
  • For in vitro use only.
  • According to ISO 6579-1/-2, ISO 21528-1, and ISO 22964, self-prepared BPW can be stored in closed containers at (5 ± 3 °C) in the dark and protected against evaporation for up to six months.

Legal Information

ECOCULT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Cell culture media expertise accelerates lab-grown meat and seafood industry with animal-free formulations.

Cell culture media expertise accelerates lab-grown meat and seafood industry with animal-free formulations.

Cell culture media expertise accelerates lab-grown meat and seafood industry with animal-free formulations.

Cell culture media expertise accelerates lab-grown meat and seafood industry with animal-free formulations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service