Skip to Content
Merck
All Photos(1)

Key Documents

1.24807

Millipore

6-(Furfurylamino)purine

for biochemistry

Synonym(s):

6-(Furfurylamino)purine, Kinetine, Furfuryladenine, Quinetine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9N5O
CAS Number:
Molecular Weight:
215.21
MDL number:
UNSPSC Code:
12352116
EC Index Number:
208-382-2
NACRES:
NA.21

Quality Level

Assay

≥99% (HPLC)

form

solid

pH

5.5-6.5 (20 °C, 10 g/L in H2O, filtered suspension)

mp

265-270 °C

bulk density

360 kg/m3

storage temp.

2-30°C

SMILES string

[nH]1c2c(nc1)ncnc2NCc3[o]ccc3

InChI

1S/C10H9N5O/c1-2-7(16-3-1)4-11-9-8-10(13-5-12-8)15-6-14-9/h1-3,5-6H,4H2,(H2,11,12,13,14,15)

InChI key

QANMHLXAZMSUEX-UHFFFAOYSA-N

Analysis Note

Assay (HPLC): ≥ 99
Identity (IR-spectrum): passes test
Heavy metals (as Pb): ≤ 0.001 %

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service