Skip to Content
Merck

0797

GVPC Agar

plate diam. 90 mm, ready-to-use, sterile, box of 20 plates, selective for Legionella spp

Synonym(s):

Glycine-vancomycin-polymyxin-cycloheximide agar, Legionella Growth Medium

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106217
NACRES:
NA.74
Plate diam.:
90 mm
Feature:
ready-to-use
Sterility:
sterile
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


agency

ISO 11731

sterility

sterile

form

plate (Semi-solid Media Filled)

feature

ready-to-use

packaging

box of 20 plates

technique(s)

microbiological culture: suitable

plate diam.

90 mm

pH

6.9 ± 0.2 (25 °C)

application(s)

food and beverages
water monitoring

storage temp.

2-8°C

suitability

selective for Legionella spp

General description

GVPC Agar, also known as Glycine-vancomycin-polymyxin-cycloheximide agar, is a Ready-to-Use media designed as per ISO Standards 11731 to detect Legionella spp.
The activated carbon in GVPC agar has an antitoxic effect due to the adsorption of fatty acids, but especially due to the breakdown of oxygen radicals and H2O2. Yeast extract provides the necessary proteins and nutrients. L-cysteine is an essential growth factor for all Legionella. The soluble iron (III) pyrophosphate [Fe4(P2O7)3] is used as a special iron source, whose quality significantly affects the growth of the Legionella. α-ketoglutarate stimulates the growth of legionella both as a carbon and nitrogen source (Krebs Cycle) as well as through mediation of the iron transport, the high amount of glycine and polymyxin B act against gram-negative, Vancomycin act against gram-positive bacteria of the accompanying bacterial flora. Cycloheximide acts fungicidal against both yeasts and molds. Thus, the selectivity of the medium is increased.

Application

Glycine Vancomycin Polymyxin Cycloheximide (GVPC) Agar is a selective culture medium used for the isolation, detection and enumeration of Legionella spp. in water samples from various sources (water intended for human consumption, wastewater, thermal and mineral waters, etc.) and food and beverage samples.


Storage Class

13 - Non Combustible Solids

WGK

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library