Skip to Content
Merck
All Photos(2)

Key Documents

07-261-I

Sigma-Aldrich

Anti-mGluR2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

07-261, Histone H2AX, H2a/x, Histone H2A.X

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

species reactivity (predicted by homology)

mouse (based on 100% sequence homology)

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Grm2(108068)
rat ... Grm2(24415)

General description

The metabotropic glutamate receptors belong to a family of G protein-coupled receptors that have been divided into 3 groups based on their sequence homology, putative signal transduction mechanisms, and pharmacologic properties. mGluR2 belongs to the Group II. It is widely expressed in different regions of fetal and the adult brain. Ligand binding to GluR2 induces a conformation change in the receptor, which triggers signaling to modulate the activity of down-stream effectors, mainly via the inhibition of adenylate cyclase activity. Activation of mGluR2 leads to inhibition of glutamate release at the synaptic junction

Immunogen

Epitope: Cytoplasmic domain
KLH-conjugated linear peptide corresponding to the cytoplasmic domain of mouse mGlur2.

Application

Anti-mGluR2 Antibody is an antibody against mGluR2 for use in Western Blotting, Immunoprecipitation.
Immunoprecipitation Analysis: 10 µg from a representative lot immunoprecipitated mGlur2 in 500 µg of rat brain microsomal prep.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blotting in rat brain microsomal prep.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected mGlur2 in 10 µg of rat brain microsomal prep.

Target description

~95/200 kDa observed. The lower band corresponds to the calculated molecular weight of mGluR2 (~100 kDa) and the upper band is likely a receptor dimer and frequently appears in immunoblots of mGluRs (Reid and Romano, 2001; Romano et al., 1996).

Linkage

Replaces: 07-261

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter U Hámor et al.
Pharmacology, biochemistry, and behavior, 175, 89-100 (2018-09-22)
Chronic methamphetamine (meth) abuse often turns into a compulsive drug-taking disorder accompanied by persistent cognitive deficits and re-occurring psychosis. Possible common neurobiological substrates underlying meth-induced deficits and schizophrenia remain poorly understood. Serotonin 2A (5-HT2A) and metabotropic glutamate 2 (mGlu2) receptors
Christian M Wood et al.
Neuropharmacology, 115, 128-138 (2016-03-19)
Modulation of metabotropic glutamate 2 (mGlu2) receptor function has huge potential for treating psychiatric and neurological diseases. Development of drugs acting on mGlu2 receptors depends on the development and use of translatable animal models of disease. We report here a
Carly N Logan et al.
Psychopharmacology, 237(7), 2007-2018 (2020-05-10)
The beta-lactam antibiotic ceftriaxone reliably attenuates the reinstatement of cocaine seeking. While the restoration of nucleus accumbens core (NA core) GLT-1 expression is necessary for ceftriaxone to attenuate reinstatement, AAV-mediated GLT-1 overexpression is not sufficient to attenuate reinstatement and does

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service