Skip to Content
Merck
All Photos(3)

Key Documents

05-623-Z

Sigma-Aldrich

Anti-RNA polymerase II Antibody, clone CTD4H8, Ascites Free

clone CTD4H8, from mouse

Synonym(s):

DNA-directed RNA polymerase II subunit RPB1, RNA polymerase II subunit B1, DNA-directed RNA polymerase II subunit A, DNA-directed RNA polymerase III largest subunit, RNA-directed RNA polymerase II subunit RPB1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

CTD4H8, monoclonal

species reactivity

mouse, yeast, rat, human

technique(s)

ChIP: suitable (ChIP-seq)
immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... POLR2A(5430)
mouse ... Polr2A(20020)
rat ... Polr2A(363633)

General description

RNA polymerase II (Pol II) is a multi-subunit enzyme responsible for the transcription of protein-coding genes. Transcription initiation requires recruitment of the complete transcription machinery to a promoter via solicitation by activators and chromatin remodeling factors. Pol II can coordinate 10 to 14 subunits. This complex interacts with the promoter regions of genes and a variety of elements and transcription factors. The DNA binding domain of the polymerase is a groove where TFIIB orients the DNA for unwinding and transcription.

Immunogen

Linear peptide corresponding to human RNA polymerase II.

Application

Anti-RNA polymerase II Antibody, clone CTD4H8, Ascites Free is a highly specific mouse monoclonal antibody, that targets RNA Polymerase & has been tested in western blotting, ICC & ChIP-seq.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
Western Blotting Analysis: 1 µg/mL from a representative lot detected RNA polymerase II in 10 µg of HeLa cell lysate.

Immunocytochemistry Analysis: A 1:50 and 1:250 dilution from a representative lot detected RNA polymerase II in HeLa and A431 cells, respectively.

Quality

Evaluated by Western Blotting in NIH/3T3 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected RNA polymerase II in 10 µg of NIH/3T3 cell lysate.

Target description

~220 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. (This product is Ascites Free.)

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bachar Dahro et al.
The New phytologist, 235(6), 2331-2349 (2022-06-14)
Invertase (INV)-mediated sucrose (Suc) hydrolysis, leading to the irreversible production of glucose (Glc) and fructose (Frc), plays an essential role in abiotic stress tolerance of plants. However, the regulatory network associated with the Suc catabolism in response to cold environment
Liang Chen et al.
Molecular cell, 68(4), 745-757 (2017-11-07)
R-loop, a three-stranded RNA/DNA structure, has been linked to induced genome instability and regulated gene expression. To enable precision analysis of R-loops in vivo, we develop an RNase-H-based approach; this reveals predominant R-loop formation near gene promoters with strong G/C skew
Double Blast Wave Primary Effect on Synaptic, Glymphatic, Myelin, Neuronal and Neurovascular Markers.
Iacono, et al.
Brain sciences, 13 (2023)
Yujing Huang et al.
Virologica Sinica, 37(3), 358-369 (2022-05-11)
Human cytomegalovirus (HCMV) is a ubiquitous pathogen belongs to betaherpesvirus subfamily. RNA2.7 is a highly conserved long non-coding RNA accounting for more than 20% of total viral transcripts. In our study, functions of HCMV RNA2.7 were investigated by comparison of
Tianxiang Hu et al.
Epigenetics, 12(3), 226-237 (2017-02-07)
LTR retrotransposons are repetitive DNA elements comprising ∼10% of the human genome. They are silenced by hypermethylation of cytosines in CpG dinucleotides and are considered parasitic DNA serving no useful function for the host genome. However, hypermethylated LTRs contain enhancer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service