Skip to Content
Merck
All Photos(1)

Key Documents

880430O

Avanti

18:1 PEG350 PE

Avanti Research - A Croda Brand 880430O

Synonym(s):

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-350] (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 25 mg (880430O-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880430O

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(OCCOCCOCCOCCOCCOCCOCCOC)=O)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[NH4+]

General description

18:0 PEG350 PE is a polyethylene glycol (PEG) conjugated phospholipid. PEGylated phospholipids are amphiphilic polymers, The molecular weight and exact mass are averages based on the polydispersity of PEG.

Application

18:1 PEG350 PE is suitable for use:
  • in the preparation of lipid bilayer membranes to estimate the diameter of the ion channel formed by the Bacillus anthracis protective antigen 63 (PA63)
  • to prepare Gd-C5F12-phospholipid nanobubbles
  • to form aqueous dispersions and to study metal cation induced cubic phase

Biochem/physiol Actions

Polyethylene glycol (PEG) conjugated phospholipids are known to protect the steric nature of liposomes. Hence, they are commonly recommended for use in liposome formulation. PEGylated lipids are used as drug carriers. Use of highly concentrated PEG-lipids makes the possibility of mixed micelles.

Packaging

5 mL Clear Glass Sealed Ampule (880430O-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Se-Young Choi et al.
PloS one, 7(3), e34333-e34333 (2012-04-06)
To develop a long-circulating theragnostics, meaning therapeutics and diagnostics for MR-guided HIFU ablation, we designed and prepared Gd-C(5)F(12)-phospholipid nanobubbles (PLNs) 30-100 nm in diameter. The biochemical and physical characterization of Gd-C(5)F(12)-PLNs were performed. Since Gd-C(5)F(12)-PLN-50 (Φ = 50 nm) and Gd-C(5)F(12)-PLN-100 (Φ = 100
Micro- and Nanoengineering of the Cell Surface (2014)
Michela Pisani et al.
The journal of physical chemistry. B, 112(17), 5276-5278 (2008-04-11)
Metal cations (Mn(2+) or Ca(2+)) in aqueous dispersions of mixtures of dioleoylphosphatidylethanolamine (DOPE) and poly(ethylene glycol)-functionalized DOPE (DOPE-PEG(350)) induce, above a certain amount of the PEG lipid component, a phase transition from the inverted hexagonal phase H(II) to the bicontinuous
Brian J Nablo et al.
Biophysical journal, 95(3), 1157-1164 (2008-07-23)
Nonelectrolyte polymers of poly(ethylene glycol) (PEG) were used to estimate the diameter of the ion channel formed by the Bacillus anthracis protective antigen 63 (PA(63)). Based on the ability of different molecular weight PEGs to partition into the pore and
G F Peacock et al.
Drug delivery, 10(1), 29-34 (2003-01-30)
A new cholesterol-carborane conjugate (BCH) has been synthesized as a potential targeting agent for boron neutron capture therapy (BNCT) of cancers. The compound is extremely water insoluble and was formulated in two liposomal formulations to determine if the compound could

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service