857395P
Avanti
GT-11
N-[(1R,2S)-2-hydroxy-1-hydroxymethyl-2-(2-tridecyl-1-cyclopropenyl)ethyl]octanamide, powder
Synonym(s):
C8-CPPC, C8-cyclopropenylceramide, GT-11 dihydroceramide desaturase inhibitor
About This Item
Recommended Products
Assay
>99% (TLC)
form
powder
packaging
pkg of 1 × 1 mg (857395P-1mg)
manufacturer/tradename
Avanti Research™ - A Croda Brand 857395P
lipid type
bioactive lipids
sphingolipids
shipped in
dry ice
storage temp.
−20°C
SMILES string
CCCCCCCCCCCCCCC1=C(C1)[C@@H](O)[C@@H](NC(CCCCCCC)=O)CO
InChI
1S/C28H53NO3/c1-3-5-7-9-10-11-12-13-14-15-17-18-20-24-22-25(24)28(32)26(23-30)29-27(31)21-19-16-8-6-4-2/h26,28,30,32H,3-23H2,1-2H3,(H,29,31)/t26-,28+/m0/s1
InChI key
AQFQOEPCXZMMDT-XTEPFMGCSA-N
General description
Biochem/physiol Actions
Packaging
Legal Information
Storage Class Code
11 - Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service