Skip to Content
Merck
All Photos(2)

Key Documents

850408C

Avanti

4ME 16:0 PS

1,2-diphytanoyl-sn-glycero-3-phospho-L-serine (sodium salt), chloroform

Synonym(s):

1,2-di-(3,7,11,15-tetramethylhexadecanoyl)-sn-glycero-3-phospho-L-serine (sodium salt); PS(16:0(3me,7me,11me,15me)/16:0(3me,7me,11me,15me))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H89NO10PNa
CAS Number:
Molecular Weight:
870.16
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (850408C-10mg)
pkg of 1 × 2.5 mL (850408C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850408C

concentration

10 mg/mL (850408C-10mg)
10 mg/mL (850408C-25mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OC[C@](C([O-])=O)([H])[NH3+])=O)(OC(CC(C)CCCC(C)CCCC(C)CCCC(C)C)=O)COC(CC(C)CCCC(C)CCCC(C)CCCC(C)C)=O.[Na+]

InChI key

KYWPZKCRBGDOKQ-YLTQECSMSA-M

Application

4ME 16:0 PS or 1,2-diphytanoyl-sn-glycero-3-phospho-L-serine (DPhPS) is suitable for use:
  • in the preparation of planar lipid bilayers
  • as an internal standard in mass spectrometry for lipidomic analyses
  • in the preparation of black lipid membranes
  • as a substrate for phaiodactylipin enzyme

Packaging

5 mL Clear Glass Sealed Ampule (850408C-10mg)
5 mL Clear Glass Sealed Ampule (850408C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A critical reassessment of penetratin translocation across lipid membranes
Barany-Wallje E, et al.
Biophysical Journal, 89(4), 2513-2521 (2005)
Phaiodactylipin, a glycosylated heterodimeric phospholipase A2 from the venom of the scorpion Anuroctonus phaiodactylus
Valdez-Cruz NA, et al.
European Journal of Biochemistry, 271(8), 1453-1464 (2004)
Does the lipid environment impact the open-state conductance of an engineered beta-barrel protein nanopore?
Tomita N, et al.
Biochimica et Biophysica Acta - Biomembranes, 1828(3), 1057-1065 (2013)
The early metabolomic response of adipose tissue during acute cold exposure in mice
Lu X, et al.
Scientific Reports, 7(1), 3455-3455 (2017)
Alessio Sacconi et al.
Langmuir : the ACS journal of surfaces and colloids, 34(40), 12183-12190 (2018-09-16)
Solid supported membranes (SSMs) are usually formed by an hybrid octadecanethiol/phosphatidylcholine (PC) bilayer supported by a gold electrode. Recently, it was shown that phosphatidylserine (PS) in place of PC can promote a more effective accumulation of lipid vesicles on the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service