Skip to Content
Merck
All Photos(2)

Key Documents

810206P

Avanti

NBD Sphinganine

omega(7-nitro-2-1,3-benzoxadiazol-4-yl)(2S,3R)-2-aminooctadecane-1,3-diol, powder

Synonym(s):

D-erythro-sphinganine-N-NBD; NBD-dihydrosphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H41N5O5
CAS Number:
Molecular Weight:
479.61
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 100 μg ((810206P-100UG))
pkg of 1 × 250 μg (810206P-250ug)
pkg of 1 × 50 μg (810206P-50UG)

manufacturer/tradename

Avanti Research - A Croda Brand 810206P

shipped in

dry ice

storage temp.

−20°C

General description

Sphinganine, also called as dihydrosphingosine is a common sphingoid base in phospholipids.

Application

NBD Sphinganine or omega (7-nitro-2-1,3-benzoxadiazol-4-yl)(2S,3R)-2-aminooctadecane-1,3-diol is suitable for the analysis of the dihydrosphingosine (DHS) localization to endoplasmic reticulum for the activation of activating transcription factor 6 (ATF6) protein.
NBD Sphinganine or omega(7-nitro-2-1,3-benzoxadiazol-4-yl)(2S,3R)-2-aminooctadecane-1,3-diol is suitable for the analysis of the dihydrosphingosine (DHS) localization to endoplasmic reticulum for the activation of activating transcription factor 6 (ATF6) protein.

Packaging

5 mL Amber Glass Screw Cap Vial (810206P-100UG)
5 mL Amber Glass Screw Cap Vial (810206P-250UG)
5ML Glass Amber Screw Cap Vial (810206P-50UG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arvin B Tam et al.
Developmental cell, 46(3), 327-343 (2018-08-08)
The unfolded protein response (UPR) is induced by proteotoxic stress of the endoplasmic reticulum (ER). Here we report that ATF6, a major mammalian UPR sensor, is also activated by specific sphingolipids, dihydrosphingosine (DHS) and dihydroceramide (DHC). Single mutations in a previously
Junliang Wan et al.
Journal of agricultural and food chemistry, 67(46), 12953-12961 (2019-10-23)
Most common sphingolipids are comprised of "typical" sphingoid bases (sphinganine, sphingosine, and structurally related compounds) and are produced via the condensation of l-serine with a fatty acyl-CoA by serine palmitoyltransferase. Some organisms, including mammals, also produce "atypical" sphingoid bases that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service