Skip to Content
Merck
All Photos(1)

Key Documents

700187P

Avanti

Galactosyl Cholesterol

Avanti Research - A Croda Brand, powder

Synonym(s):

β-D-Galactosyl cholesterol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H56O6
CAS Number:
Molecular Weight:
548.79
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 100 mg (700187P-100MG)
pkg of 1 × 5 mg (700187P-5MG)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

C[C@H](CCCC(C)C)[C@H]1CCC2C3CC=C4C[C@@H](O[C@@H]5O[C@H](CO)[C@H](O)[C@H](O)[C@H]5O)CC[C@]4(C)C3CC[C@]12C

InChI

1S/C33H56O6/c1-19(2)7-6-8-20(3)24-11-12-25-23-10-9-21-17-22(13-15-32(21,4)26(23)14-16-33(24,25)5)38-31-30(37)29(36)28(35)27(18-34)39-31/h9,19-20,22-31,34-37H,6-8,10-18H2,1-5H3/t20-,22+,23?,24-,25?,26?,27-,28+,29+,30-,31-,32+,33-/m1/s1

InChI key

FSMCJUNYLQOAIM-CHGMBOCPSA-N

Biochem/physiol Actions

Galactosyl Cholesterol (GalChol) is used in the preparation of vesicles. The vesicles formed with GalChol is more stable in vivo and have higher half-life period. GalChol aggregates becomes larger in chloroform. GalChol has lower activation energy and do not form complexes in bilayer plane. Borrelia sp. possesses GalChol, binds cholesterol and a molecule of D-galactose in the place of D-glucose.

Packaging

5 mL Amber Glass Screw Cap Vial (700187P-5MG and 700187P-100MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steroids (2011)
Stability of carbohydrate-modified vesicles in vivo: comparative effects of ceramide and cholesterol glycoconjugates.
Wu PS, et al.
Proceedings of the National Academy of Sciences of the USA, 79(18), 5490-5493 (1982)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service