Key Documents
Z517445
Sigma-Aldrich
Aldrich® triple-jacket coil condenser
joint: ST/NS 29/32
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Joint
joint: ST/NS29/32
O.D.
60 mm
surface area
1230 cm2
General description
Excellent for lower boiling solvents.
Features:
• Triple-jacketing provides maximum cooling surface area
• Does not flood as readily during reflux surges
• Minimizes loss of volatile components
• Highly efficient while extremely compact
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)