Skip to Content
Merck
All Photos(1)

Key Documents

W328655

Sigma-Aldrich

Tripropionin

≥97.1%, FCC, FG

Synonym(s):

Glyceryl tripropionate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5CO2CH2)2CH(O2CC2H5)
CAS Number:
Molecular Weight:
260.28
FEMA Number:
3286
MDL number:
UNSPSC Code:
12164502
Flavis number:
9.263
NACRES:
NA.21
grade:
FG
Halal
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117

vapor density

8.95 (vs air)

Assay

≥97.1%

autoignition temp.

790 °F

expl. lim.

0.8 %, 186 °F

refractive index

n20/D 1.434 (lit.)

bp

175-176 °C/20 mmHg (lit.)

density

1.08 g/mL at 25 °C (lit.)

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

oily; chemical

SMILES string

CCC(=O)OCC(COC(=O)CC)OC(=O)CC

InChI

1S/C12H20O6/c1-4-10(13)16-7-9(18-12(15)6-3)8-17-11(14)5-2/h9H,4-8H2,1-3H3

InChI key

YZWRNSARCRTXDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • The gut microbiome modulates the impact of Anaerobutyricum soehngenii supplementation on glucose homeostasis in mice.: This study investigates how the gut microbiome affects glucose homeostasis in mice supplemented with Anaerobutyricum soehngenii. Findings suggest a significant role of microbial interactions in metabolic regulation (Hutchison et al., 2024).

  • Development and cytotoxicity evaluation of multiple nanoemulsions for oral co-delivery of 5-fluorouracil and short chain triglycerides for colorectal cancer.: This paper explores the development of nanoemulsions containing 5-fluorouracil and short-chain triglycerides, including tripropionin, for targeted cancer therapy, highlighting their potential in improving drug delivery and efficacy (Fukumori et al., 2023).

  • Gastrointestinal digestion of Pickering emulsions stabilised by hydrophobically modified cellulose nanocrystals: Release of short-chain fatty acids.: This study examines the digestion of Pickering emulsions stabilized by modified cellulose nanocrystals, focusing on the release of short-chain fatty acids, including those from tripropionin, during gastrointestinal digestion (Le et al., 2020).

  • Evaluation of lipid-stabilised tripropionin nanodroplets as a delivery route for combretastatin A4.: This research evaluates the use of lipid-stabilized tripropionin nanodroplets for the delivery of the anticancer drug combretastatin A4, demonstrating enhanced stability and targeted delivery potential (Mico et al., 2017).

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service