Key Documents
SYNC159200
Synthware West condenser
joint: ST/NS 19/22, jacket L 200 mm
Synonym(s):
Synthware C159200, glass West condenser
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Joint
joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C159200
jacket L
200 mm
Related Categories
General description
With narrow annular space for fast coolant speed, thus high cooling efficiency.
Legal Information
Synthware is a trademark of Kemtech America Inc.