S6452
Succinyl chloride
95%
Synonym(s):
Succinic acid dichloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
ClCOCH2CH2COCl
CAS Number:
Molecular Weight:
154.98
Beilstein:
507064
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Quality Level
Assay
95%
refractive index
n20/D 1.468 (lit.)
bp
190 °C (lit.)
95-96 °C/16 mmHg (lit.)
mp
15-18 °C (lit.)
density
1.407 g/mL at 25 °C (lit.)
SMILES string
ClC(=O)CCC(Cl)=O
InChI
1S/C4H4Cl2O2/c5-3(7)1-2-4(6)8/h1-2H2
InChI key
IRXBNHGNHKNOJI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Supplementary Hazards
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
168.8 °F - closed cup
Flash Point(C)
76 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Afrasim Moin et al.
Current drug delivery, 13(7), 1034-1052 (2016-05-04)
The management of TB is difficult due to the resistance developed by Mycobacterium tuberculosis against anti-tubercular drugs causes the new challenges for treating and managing the disease. The objective of the study was to combat multiple drug resistance (MDR) exhibited
Florian Diot-Néant et al.
ChemSusChem, 13(10), 2613-2620 (2020-04-03)
The already-reported, low-yielding, and non-sustainable Et3 N-mediated homocoupling of levoglucosenone (LGO) into the corresponding LGO-CyreneTM diketone has been revisited and greened-up. The use of methanol as both a renewable solvent and catalyst and K2 CO3 as a safe inorganic base
Silvio Curia et al.
ChemSusChem, 11(15), 2529-2539 (2018-06-21)
The utilization of wood-derived building blocks (xylochemicals) to replace fossil-based precursors is an attractive research subject of modern polymer science. Here, we demonstrate that bisguaiacol (BG), a lignin-derived bisphenol analogue, can be used to prepare biobased polyesters with remarkable thermal
Na Sun et al.
International journal of molecular medicine, 45(3), 919-930 (2020-01-28)
3,4‑Dihydroxyacetophenone (DHAP) exerts therapeutic effects on cardiovascular disease and pulmonary heart disease. However, it is not utilized as a clinical drug due to its rapid metabolism, short‑acting effect and low oral bioavailability. In the present study, three derivatives of DHAP
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service