Skip to Content
Merck
All Photos(1)

Key Documents

QBD11303

Sigma-Aldrich

MAL-dPEG ®24-amido-dPEG ®24-TFP ester

Synonym(s):

Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C115H209F4N3O54
Molecular Weight:
2573.87
UNSPSC Code:
51171641
NACRES:
NA.22

Assay

>90%

form

solid or viscous liquid

reaction suitability

reactivity: thiol reactive
reagent type: cross-linking reagent
reactivity: amine reactive

polymer architecture

shape: linear
functionality: heterobifunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(CCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCNC(CCN1C(C=CC1=O)=O)=O)NCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCC(OC2=C(C(F)=CC(F)=C2F)F)=O

Features and Benefits

MAL-dPEG ®₂₄-amido-dPEG ®₂₄-TFP ester can be used to conjugate two different proteins or a protein with a payload by reaction of the maleimide with a free thiol and reaction of the TFP ester with a primary amine.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service