MT51343204
Mettler Toledo InLab® Redox Au ORP electrode
Redox Electrode with Gold Ring Indicator
All Photos(1)
About This Item
UNSPSC Code:
39121436
NACRES:
NB.79
Recommended Products
material
glass shaft
description
S7 connection
Without cable
manufacturer/tradename
Mettler-Toledo 51343204
temperature measuring range
0-100 °C
L × W
120 mm × 12 mm
General description
Mettler Toledo InLab® Redox Au is a combination electrode for measuring the oxidation/reduction power of a solution. Consists of a gold ring indicator electrode and a silver/silver chloride reference electrode.
The measuring signal is produced on the surface of the precious metal by an exchange of electrons with the oxidation-reduction system of the measured medium.
The combination design avoids use of separate reference and indicating electrodes.
The InLab® Redox Au electrode is fitted with the innovative Safelock closing ring which gives easy access for re-filling reference electrolyte and complete sealing when closed.
Different cables can be connected to the S7 screw head allowing use with a wide range of meters.
The measuring signal is produced on the surface of the precious metal by an exchange of electrons with the oxidation-reduction system of the measured medium.
The combination design avoids use of separate reference and indicating electrodes.
The InLab® Redox Au electrode is fitted with the innovative Safelock closing ring which gives easy access for re-filling reference electrolyte and complete sealing when closed.
Different cables can be connected to the S7 screw head allowing use with a wide range of meters.
- Combination design
- Safelock closing ring
- Detachable cable
Legal Information
InLab is a registered trademark of Mettler-Toledo, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)