Skip to Content
Merck
All Photos(1)

Key Documents

JKA5021

Sigma-Aldrich

Acrylate-PEG-NHS

average MN 2,000, cross-linking reagent polymerization reactions amine reactive

Synonym(s):

Acrylate-PEG-NHS, Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2CHCOO(CH2CH2O)nCH2COOC4H4O2N
UNSPSC Code:
12162002
NACRES:
NA.23

Product Name

Acrylate-PEG2K-NHS, average Mn 2,000

form

solid

mol wt

average Mn 2,000

reaction suitability

reagent type: cross-linking reagent
reaction type: Polymerization Reactions
reactivity: amine reactive

polymer architecture

shape: linear
functionality: heterobifunctional

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Acrylate PEG NHS Ester. Light sensitive PEG, will crosslink with exposure to ultraviolet light.

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dang, L.T.H., et al.
Biomaterials, 35(27) (2014)
Hoffman, M.D., et al.
Acta Biomaterialia, 10(8) (2014)
Qi, H., et al.
Nature Communications, 4 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service