Skip to Content
Merck
All Photos(1)

Key Documents

GF64040239

Glass Sealing Alloy, Fe54Ni29Co17

100mm tube, 6.35mm outside diameter, 0.25mm wall thickness, 5.85mm inside diameter, as drawn

Synonym(s):

FE047650

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101712
NACRES:
NA.23

form

tubes

manufacturer/tradename

Goodfellow 640-402-39

L × wall thickness

100 mm × 0.25 mm

O.D. × I.D.

6.35 mm × 5.85 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service