DWK213015409
Duran® Evaporating Dish
cylinder capacity 600 mL
Synonym(s):
213015409
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
description
With Spout
packaging
pkg of 10 ea
manufacturer/tradename
DURAN® 213015409
cylinder capacity
600 mL
Looking for similar products? Visit Product Comparison Guide
Application
Evaporation trays are used to evaporate excess water - or another solvent - to leave a concentrated solution or the substance dissolved in it.
Features and Benefits
- Spout for clean pouring
- Easy-to-read scale with graduation marks to estimate the content
- Fired-on, highly durable white ceramic
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service