Key Documents
DWK11-5200-S
Wheaton® Microliter Vial
About This Item
UNSPSC Code:
41122001
NACRES:
NB.11
material
clear vial (Silanized, with Marking Patch)
packaging
pkg of 100 ea
manufacturer/tradename
WHEATON® 11-5200-S
D × H
12 mm × 32 mm
cap size
11 mm
volume
2.0 mL
Application
Chromatography
Features and Benefits
  • Double ring finish for added closure performance.
  • Approved for LC & GC.
  • Where noted, Certified Class 10,000 clean room packed enhances data integrity.
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)