D1203
1,10-Decanediol
98%
Synonym(s):
Decamethylene glycol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HO(CH2)10OH
CAS Number:
Molecular Weight:
174.28
Beilstein:
1698975
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
form
solid
SMILES string
OCCCCCCCCCCO
InChI
1S/C10H22O2/c11-9-7-5-3-1-2-4-6-8-10-12/h11-12H,1-10H2
InChI key
FOTKYAAJKYLFFN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
305.6 °F - closed cup
Flash Point(C)
152 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
J Helle et al.
Biomaterials, 23(22), 4397-4404 (2002-09-11)
Poly(ortho esters) with a low glass transition temperature are semi-solid materials so that therapeutic agents can be incorporated at room temperature, without the use of solvents, by a simple mixing procedure. When molecular weights are limited to < 5 kDa
O A Vinogradova et al.
Bioorganicheskaia khimiia, 35(3), 384-396 (2009-07-22)
An opportunity of designing nontypical double-stranded DNA structures containing nonnatural inserts in a regular nucleotide DNA sequence has been investigated. The looped nucleotide inserts on the basis of adenylates and thymidilates, and nonnucleotide inserts on the basis of phosphodiesters of
K Schwach-Abdellaoui et al.
Journal of biomaterials science. Polymer edition, 10(3), 375-389 (1999-04-03)
The synthesis of poly(ortho-esters) (POEs) containing lactic acid dimers in the polymer backbone for possible use in controlled drug release applications is described. These autocatalyzed POEs are prepared by the acid catalyzed condensation of 3,9-diethylidene-2,4,8,10-tetraoxaspiro(5,5)undecane with diols to produce linear
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service