All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C14H7ClO2
CAS Number:
Molecular Weight:
242.66
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
powder
mp
159-160 °C (lit.)
SMILES string
Clc1cccc2C(=O)c3ccccc3C(=O)c12
InChI
1S/C14H7ClO2/c15-11-7-3-6-10-12(11)14(17)9-5-2-1-4-8(9)13(10)16/h1-7H
InChI key
BOCJQSFSGAZAPQ-UHFFFAOYSA-N
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Yue Fan et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 6(1), 1801368-1801368 (2019-01-16)
Disperse reactive dyes with appropriate chemical structure are key for the coloration of natural fibers in the water-free environmentally friendly medium of supercritical carbon dioxide with various advantages. The objective of this work is to design and synthesize a novel
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service