BR87604
BRAND® PP beaker with spout, low form
volume 25 mL
Synonym(s):
Beakers, Plastics
All Photos(3)
About This Item
Recommended Products
material
polypropylene
description
ISO 7056
feature
graduations
beaded rim and spout
manufacturer/tradename
BRAND 87604
volume
25 mL
General description
The BRAND® PP Beaker with Spout in low form is crafted from high-clarity polypropylene (PP) and complies with ISO 7056 standards, ensuring durability and reliability for laboratory applications. This beaker features clear graduation markings for precise measurement, along with a beaded rim that enhances strength and provides a comfortable grip. The integrated spout allows for easy pouring, minimizing spills and improving accuracy during liquid transfer. To preserve the marks and inscriptions, it is recommended to clean the beaker at temperatures up to a maximum of 60 °C.
Other Notes
Browse all plastic beakers
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG