Key Documents
BR81265
BRAND® spare key for automatic burettes Pellet pattern
PTFE key
Synonym(s):
Accessories, Burette stopcock, Burettes, Compact Burettes, Stopcocks
About This Item
UNSPSC Code:
41121808
NACRES:
NB.43
material
PTFE key
packaging
pack of 1 ea
manufacturer/tradename
BRAND 81265
General description
The BRAND® spare key for automatic burettes features a pellet pattern design and is made from durable PTFE, ensuring chemical resistance and longevity. This spare key is equipped with a retention device, providing secure attachment and preventing accidental loss during use. It is specifically designed to enhance the functionality of automatic burettes, allowing for smooth and precise liquid dispensing.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)