Skip to Content
Merck
All Photos(1)

Documents

BR148045

BRAND® PP powder funnel short wide stem

top diam. 180 mm, bottom diam. 40 mm

Synonym(s):

Funnels, Powder funnels

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121809
NACRES:
NB.43

packaging

pack of 1 ea

manufacturer/tradename

BRAND 148045

bottom diam.

40 mm

top diam.

180 mm

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • BRAND® powder funnel is ideal for transfering powders.
  • Made from break-resistant polypropylene, these general lab use funnels include a short wide stem that facilitates optimal filtration.

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service