Skip to Content
Merck
All Photos(3)

Documents

ALD00236

Sigma-Aldrich

Sodium N-Boc-azetidine-3-sulfinate

Synonym(s):

Sodium 1-(tert-butoxycarbonyl)azetidine-3-sulfinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14NNaO4S
Molecular Weight:
243.26
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

powder

Quality Level

reaction suitability

reaction type: C-C Bond Formation

mp

172-177 °C (w/ decomp)

storage temp.

2-8°C

SMILES string

O=S([O-])C1CN(C(OC(C)(C)C)=O)C1.[Na+]

InChI

1S/C8H15NO4S.Na/c1-8(2,3)13-7(10)9-4-6(5-9)14(11)12;/h6H,4-5H2,1-3H3,(H,11,12);/q;+1/p-1

InChI key

KYPHAOQWDKDUAC-UHFFFAOYSA-M

Application

The following Baran Sulfinate is a part of a toolbox of diversification reagents, which are ideal for the late-stage functionalization of nitrogen-containing heterocycles. The regioselectivity can be effectively tuned by modification of the pH and solvent selection.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service