Skip to Content
Merck
All Photos(1)

Key Documents

933066

Sigma-Aldrich

Tweezers

Style 3F

Synonym(s):

Tweezer, Grippers, Tweezers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111761
NACRES:
NB.35

Looking for similar products? Visit Product Comparison Guide

General description

EMS techware

Application

High precision and ultrafine tweezers. Typical applications include tweezers for the electronic industry, watch-makers, jewelers and laboratory in moderately aggressive chemical environments. Tips touching over 3mm (parallel tips), useful for handling tiny wires. Length: 120mm.
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish. Tips touching over 3mm (parallel tips). Useful for handling tiny wires.
Material: SA Steel. OAL: 120 mm. Tips: Parallel touching.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service