Skip to Content
Merck
All Photos(1)

Key Documents

918768

Sigma-Aldrich

PIM-1

Synonym(s):

2,3,5,6-Tetrafluorophthalonitrile-3,3,3′,3′-tetramethyl-1,1′-spirobisin dane-5,5′,6,6′-tetrol co-polymer, Polymers of intrinsic microporosity

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C29H20N2O4)n
UNSPSC Code:
12162002
NACRES:
NA.23

Quality Level

Application

Polymers of intrinsic microporosity (PIMs) have generated significant interest for membrane separations and other applications since their initial discovery in 2004. PIMs are designed to have highly rigid and contorted backbone structures that cannot pack efficiently and thus exhibit microporosity. PIMs find applications in industry as they are solution processable, have a high permeability, and exhibit significant gas pair selectivities as membranes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polymers of Intrinsic Microporosity
Neil B.
ISRN Materials Science, 2012, 16-16 null
Understanding the Topology of the Polymer of Intrinsic Microporosity PIM-1: Cyclics, Tadpoles, and Network Structures and Their Impact on Membrane Performance.
Foster A B, et al.
Macromolecules, 53(2), 569-583 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service