Skip to Content
Merck
All Photos(2)

Key Documents

901000

Sigma-Aldrich

1,4-Cubanedicarboxylic acid

≥97%

Synonym(s):

(1S,2R,3R,4S,5R,6S,7S,8S)-Cubane-1,4-dicarboxylic acid, Pentacyclo[4.2.0.02,5.03,8.04,7]octane-1,4-dicarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8O4
CAS Number:
Molecular Weight:
192.17
UNSPSC Code:
12352106
NACRES:
NA.22

Quality Level

Assay

≥97%

form

powder

mp

224 °C

SMILES string

OC([C@]12[C@H]3[C@H]4[C@@H]1[C@H]5[C@@]4(C(O)=O)[C@H]3[C@@H]52)=O

InChI

1S/C10H8O4/c11-7(12)9-1-2-4(9)6-5(9)3(1)10(2,6)8(13)14/h1-6H,(H,11,12)(H,13,14)

InChI key

JFKXMJUMOJJTCQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

1,4-Cubanedicarboxylic acid can undergo esterification with alkylsulfuric acids. The acid and its ester derivatives are also capable of undergoing fluorination with elemental fluorine. This product was previously listed as BML00135.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service