Skip to Content
Merck
All Photos(1)

Key Documents

743011

Sigma-Aldrich

O-(2-Mercaptoethyl)-O′-methylpolyethylene glycol

10,000

Synonym(s):

Polyethylene glycol, MeO-PEG-SH, Mercaptopolyethylene glycol monomethyl ether, PEG-thiol, Polyethylene glycol 2-mercaptoethyl methyl ether

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
51171641
PubChem Substance ID:
NACRES:
NA.22

Quality Level

form

lumps

mol wt

average Mw 10,000

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service