666939
3-Bromo-N,N-dimethylaniline
97%
Synonym(s):
(3-Bromophenyl)dimethylamine, 1-Bromo-3-dimethylaminobenzene, 3-(N,N-Dimethylamino)-1-bromobenzene, 3-Bromo-N,N-dimethylaniline, 3-Bromo-N,N-dimethylbenzenamine, N-(3-Bromophenyl)dimethylamine
About This Item
Recommended Products
Assay
97%
refractive index
n20/D 1.6015
density
1.402 g/mL at 25 °C
functional group
amine
bromo
SMILES string
CN(C)c1cccc(Br)c1
InChI
1S/C8H10BrN/c1-10(2)8-5-3-4-7(9)6-8/h3-6H,1-2H3
InChI key
USEXQPWLCGBYNT-UHFFFAOYSA-N
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2 - STOT RE 2
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service