608556
Deuterium oxide-18O
5 atom % D, 5 atom % 18O
Synonym(s):
Doubly Labeled Water, Heavy water-d2,18O, Unnormalized water-18O
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
D218O
CAS Number:
Molecular Weight:
22.03
MDL number:
UNSPSC Code:
12352303
Recommended Products
isotopic purity
5 atom % 18O
5 atom % D
bp
101.4 °C (lit.)
mp
3.8 °C (lit.)
mass shift
M+4
SMILES string
[2H][18O][2H]
InChI
1S/H2O/h1H2/i1+2/hD2
InChI key
XLYOFNOQVPJJNP-LAPWFLRPSA-N
Looking for similar products? Visit Product Comparison Guide
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service