603724
2-Iodopropane-1-13C
≥99 atom % 13C, ≥99% (CP), contains copper as stabilizer
Synonym(s):
Isopropyl iodide-1-13C
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3CHI13CH3
CAS Number:
Molecular Weight:
170.98
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
Recommended Products
isotopic purity
≥99 atom % 13C
Assay
≥99% (CP)
contains
copper as stabilizer
bp
88-90 °C (lit.)
density
1.713 g/mL at 25 °C
mass shift
M+1
SMILES string
CC([13CH3])I
InChI
1S/C3H7I/c1-3(2)4/h3H,1-2H3/i1+1
InChI key
FMKOJHQHASLBPH-OUBTZVSYSA-N
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
107.6 °F - closed cup
Flash Point(C)
42 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service