Skip to Content
Merck
All Photos(1)

Key Documents

601764

Sigma-Aldrich

Argon-36Ar

50 atom %

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
36Ar
CAS Number:
Molecular Weight:
35.97
MDL number:
UNSPSC Code:
12142004
PubChem Substance ID:

isotopic purity

50 atom %

Assay

≥99.5% (Total Argon)

form

gas

impurities

<0.5% Total others

mass shift

M-4

SMILES string

[36Ar]

InChI

1S/Ar/i1-4

InChI key

XKRFYHLGVUSROY-AHCXROLUSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service