Skip to Content
Merck
All Photos(1)

Key Documents

601403

Sigma-Aldrich

Lithium-6Li chloride

95 atom % 6Li, 99% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
6LiCl
CAS Number:
Molecular Weight:
41.47
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

isotopic purity

95 atom % 6Li

Assay

99% (CP)

form

solid

mp

605 °C (lit.)

mass shift

M-1

SMILES string

[6Li+].[Cl-]

InChI

1S/ClH.Li/h1H;/q;+1/p-1/i;1-1

InChI key

KWGKDLIKAYFUFQ-HCMAANCNSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service