All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H14O3
CAS Number:
Molecular Weight:
182.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
form
solid
mp
67-70 °C (lit.)
functional group
anhydride
ester
SMILES string
O=C1CC2(CCCCC2)CC(=O)O1
InChI
1S/C10H14O3/c11-8-6-10(7-9(12)13-8)4-2-1-3-5-10/h1-7H2
InChI key
XNDSIASQMRYFSW-UHFFFAOYSA-N
Application
3-Oxaspiro[5,5]undecane-2,4-dione may be used to synthesize [1-(2-{[4-chloro-2-(methoxycarbonyl)phenyl]amino}-2-oxoethyl)cyclohexyl]acetic acid.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Nagahisa Yamaoka et al.
Chemical & pharmaceutical bulletin, 59(2), 215-224 (2011-02-08)
Novel anthranilic acid derivatives having substituted N-acyl side chains were designed and synthesized for evaluation as plasminogen activator inhibitor-1 (PAI-1) inhibitors. Compounds with a 4-diphenylmethyl-1-piperazinyl moiety on the acyl side chains in general exhibited potent in vitro PAI-1 inhibitory activity
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service