Skip to Content
Merck
All Photos(1)

Key Documents

530840

Sigma-Aldrich

Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate

98%

Synonym(s):

Bis(1,5-cyclooctadiene)rhodium(I) triflate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H24F3O3RhS
CAS Number:
Molecular Weight:
468.34
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

reaction suitability

core: rhodium
reagent type: catalyst

mp

159 °C (dec. literature value) (lit.)

SMILES string

[Rh+].[O-]S(=O)(=O)C(F)(F)F.C1CC=CCCC=C1.C2CC=CCCC=C2

InChI

1S/2C8H12.CHF3O3S.Rh/c2*1-2-4-6-8-7-5-3-1;2-1(3,4)8(5,6)7;/h2*1-2,7-8H,3-6H2;(H,5,6,7);/q;;;+1/p-1/b2*2-1-,8-7-;;

InChI key

GOPBTCMAUMSOBX-QMDOQEJBSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service