Skip to Content
Merck
All Photos(1)

Documents

469475

Sigma-Aldrich

(R)-(+)-3-(Benzyloxycarbonyl)-4-oxazolidinecarboxylic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13NO5
CAS Number:
Molecular Weight:
251.24
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

optical activity

[α]20/D +92°, c = 1 in chloroform

optical purity

ee: 97% (HPLC)

mp

70-74 °C (lit.)

SMILES string

OC(=O)[C@H]1COCN1C(=O)OCc2ccccc2

InChI

1S/C12H13NO5/c14-11(15)10-7-17-8-13(10)12(16)18-6-9-4-2-1-3-5-9/h1-5,10H,6-8H2,(H,14,15)/t10-/m1/s1

InChI key

XRRRGBIMHQARMF-SNVBAGLBSA-N

Application

Useful chiral synthon (derived from serine) for the asymmetric synthesis of a variety of nitrogen-containing targets.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Falorni, M. et al.
Tetrahedron Asymmetry, 6, 287-287 (1995)

Articles

Proline analogues are promising candidates for tuning the biological, pharmaceutical, or physicochemical properties of naturally occuring, as well as de novo designed, linear, and, cyclic peptides.

Proline analogues are promising candidates for tuning the biological, pharmaceutical, or physicochemical properties of naturally occuring, as well as de novo designed, linear, and, cyclic peptides.

Proline analogues are promising candidates for tuning the biological, pharmaceutical, or physicochemical properties of naturally occuring, as well as de novo designed, linear, and, cyclic peptides.

Proline analogues are promising candidates for tuning the biological, pharmaceutical, or physicochemical properties of naturally occuring, as well as de novo designed, linear, and, cyclic peptides.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service