Skip to Content
Merck
All Photos(1)

Documents

281859

Sigma-Aldrich

Diethyl malonate-2-13C

99 atom % 13C

Synonym(s):

1,3-Diethyl ester propanedioic-2-13C acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CH2(CO2C2H5)2
CAS Number:
Molecular Weight:
161.16
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.12

vapor density

5.52 (vs air)

Quality Level

vapor pressure

1 mmHg ( 40 °C)

isotopic purity

99 atom % 13C

Assay

99% (CP)

form

liquid

bp

199 °C (lit.)

mp

-51--50 °C (lit.)

density

1.054 g/mL at 25 °C

mass shift

M+1

SMILES string

CCOC(=O)[13CH2]C(=O)OCC

InChI

1S/C7H12O4/c1-3-10-6(8)5-7(9)11-4-2/h3-5H2,1-2H3/i5+1

InChI key

IYXGSMUGOJNHAZ-HOSYLAQJSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90.00 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service