Skip to Content
Merck
All Photos(1)

Documents

257931

Sigma-Aldrich

3-Methyl-1-butene

95%

Synonym(s):

α-Isoamylene, 2-Methyl-3-butene, 3,3-Dimethylpropene, 3-Methyl-1-butene, Isopropylethene, Isopropylethylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH=CH2
CAS Number:
Molecular Weight:
70.13
Beilstein:
505980
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

14.97 psi ( 20 °C)

Quality Level

Assay

95%

bp

20 °C (lit.)

mp

−168.5-−168 °C (lit.)

density

0.627 g/mL at 25 °C (lit.)

SMILES string

CC(C)C=C

InChI

1S/C5H10/c1-4-5(2)3/h4-5H,1H2,2-3H3

InChI key

YHQXBTXEYZIYOV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Gas-phase reaction of the OH radical with 3-methyl-1-butene in the presence of NO has been investigated. Polymerization of 3-methyl-1-butene in the presence of homogeneous metallocene catalyst (co-catalyst methylalumoxane) has been investigated.

Application

3-Methyl-1-butene has been used in asymmetric total synthesis of (−)-Linderol A, a potent inhibitor of melanin biosynthesis of cultured B-16 melanoma cells.

Recommended products

Brass hose adapter Z146811 or brass body mini gas regulator Z513539 is recommended.

hose barb

Product No.
Description
Pricing

regulator

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-70.6 °F - closed cup

Flash Point(C)

-57 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Products of the gas-phase reaction of the OH radical with 3-methyl-1-butene in the presence of NO.
Atkinson R, et al.
International Journal of Chemical Kinetics, 30(8), 577-587 (1998)
Masayuki Yamashita et al.
The Journal of organic chemistry, 72(15), 5697-5703 (2007-06-21)
The first asymmetric total synthesis of (-)-Linderol A, a potent inhibitor of melanin biosynthesis of cultured B-16 melanoma cells, has been achieved via two key reactions: a diastereoselective [2+2] photocycloaddition of a coumarin-3-carboxylate bearing a chiral auxiliary with 3-methyl-1-butene and
Polymerization of 3-methyl-1-butene promoted by metallocene catalysts.
Borriello A, et al.
Macromolecular Rapid Communications, 17(8), 589-597 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service