Skip to Content
Merck
All Photos(2)

Key Documents

237442

Sigma-Aldrich

(R)-(+)-1-(1-Naphthyl)ethylamine

≥99%

Synonym(s):

(R)-(+)-α-Methyl-1-naphthalenemethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7CH(CH3)NH2
CAS Number:
Molecular Weight:
171.24
Beilstein:
2208025
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99%

form

liquid

optical activity

[α]20/D +55°, c = 2 in ethanol

optical purity

ee: 98% (GLC)

refractive index

n20/D 1.623 (lit.)

bp

153 °C/11 mmHg (lit.)

density

1.067 g/mL at 20 °C (lit.)

SMILES string

C[C@@H](N)c1cccc2ccccc12

InChI

1S/C12H13N/c1-9(13)11-8-4-6-10-5-2-3-7-12(10)11/h2-9H,13H2,1H3/t9-/m1/s1

InChI key

RTCUCQWIICFPOD-SECBINFHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Absolute configuration of primary amines determined using chiral (2-nitrophenyl)proline amides and 1H NMR.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hee Choon Ahn et al.
Organic letters, 9(19), 3853-3855 (2007-08-23)
N-(2-Nitrophenyl)proline (2-NPP) amides of primary amines have a conformational preference for intramolecular hydrogen bonding. Because of the strong and selective anisotropic effects on the amine substituents, the absolute configuration of alpha-chiral primary amines can be assigned by comparing the 1H

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service