Merck
A quantitative model for dermal infection and oedema in BALB/c mice pinna.
BMC microbiology (2016-12-14)
Erika Nahomy Marino-Marmolejo, Flor Yohana Flores-Hernández, Mario Alberto Flores-Valdez, Luis Felipe García-Morales, Ana Cecilia González-Villegas, Jorge Bravo-Madrigal
ABSTRACT
Pharmaceutical industry demands innovation for developing new molecules to improve effectiveness and safety of therapeutic medicines. Preclinical assays are the first tests performed to evaluate new therapeutic molecules using animal models. Currently, there are several models for evaluation of treatments, for dermal oedema or infection. However, the most common or usual way is to induce the inflammation with chemical substances instead of infectious agents. On the other hand, this kind of models require the implementation of histological techniques and the interpretation of pathologies to verify the effectiveness of the therapy under assessment. This work was focused on developing a quantitative model of infection and oedema in mouse pinna. The infection was achieved with a strain of Streptococcus pyogenes that was inoculated in an injury induced at the auricle of BALB/c mice, the induced oedema was recorded by measuring the ear thickness with a digital micrometer and histopathological analysis was performed to verify the damage. The presence of S. pyogenes at the infection site was determined every day by culture. Our results showed that S. pyogenes can infect the mouse pinna and that it can be recovered at least for up to 4 days from the infected site; we also found that S. pyogenes can induce a bigger oedema than the PBS-treated control for at least 7 days; our results were validated with an antibacterial and anti-inflammatory formulation made with ciprofloxacin and hydrocortisone. The model we developed led us to emulate a dermal infection and allowed us to objectively evaluate the increase or decrease of the oedema by measuring the thickness of the ear pinna, and to determine the presence of the pathogen in the infection site. We consider that the model could be useful for assessment of new anti-inflammatory or antibacterial therapies for dermal infections.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
Terreno essenziale minimo di Eagle, With Earle′s salts, L-glutamine, and non-essential amino acids, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Wright Stain, Modified